Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 11-Maleimidoundecanoate 98.0+%, TCI America™
SDP

Catalog No. S088220MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S088220MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S088220MG Supplier TCI America Supplier No. S088220MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

CAS 87981-04-2
Melting Point 92°C
Color White
Molecular Formula C19H26N2O6
Quantity 20 mg
MDL Number MFCD00467873
Synonym 11-Maleimidoundecanoic Acid N-Succinimidyl Ester, N-(11-Maleimidoundecanoyloxy)succinimide, KMUS
InChI Key SGNVTRHWJGTNKV-UHFFFAOYSA-N
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCCCCCCCN2C(=O)C=CC2=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 11-(2,5-dioxopyrrol-1-yl)undecanoate
Molecular Weight (g/mol) 378.425
PubChem CID 5091658
Formula Weight 378.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 11-Maleimidoundecanoate
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.