accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNN

CAS RN 879896-40-9

IUPAC Name: methyl[(6-phenylpyridin-3-yl)methyl]amine
Synonyms: methyl[(6-phenylpyridin-3-yl)methyl]amine
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: OLZSHISXLYLLBI-UHFFFAOYSA-N
SMILES: CNCC1=CN=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results