accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNNN

CAS RN 879896-43-2

IUPAC Name: methyl({[3-(1H-1,2,4-triazol-1-yl)phenyl]methyl})amine
Synonyms: methyl({[3-(1,2,4-triazol-1-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 188.23
Molecular Formula: C10H12N4
InChi Key: LLGQTEDUTKNYIP-UHFFFAOYSA-N
SMILES: CNCC1=CC(=CC=C1)N1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results