accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH2N

CAS RN 879896-50-1

IUPAC Name: 1-{2-[(4-methylpiperazin-1-yl)methyl]phenyl}methanamine
Synonyms: 1-{2-[(4-methylpiperazin-1-yl)methyl]phenyl}methanamine
Molecular Weight (g/mol): 219.33
Molecular Formula: C13H21N3
InChi Key: OKGWICVOLZFMPE-UHFFFAOYSA-N
SMILES: CN1CCN(CC2=CC=CC=C2CN)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results