accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ClS

CAS RN 88-11-9

IUPAC Name: (chloromethanethioyl)diethylamine
Synonyms: (chloromethanethioyl)diethylamine
Molecular Weight (g/mol): 151.65
Molecular Formula: C5H10ClNS
InChi Key: HUUSTUALCPTCGJ-UHFFFAOYSA-N
SMILES: CCN(CC)C(Cl)=S
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)