accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88-18-6

IUPAC Name: 2-tert-butylphenol
Synonyms: 2-tert-butylphenol O-T-butylphenol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: WJQOZHYUIDYNHM-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=CC=C1O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)