Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-tert-Butylphenol

Catalog No. B9940550ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B9940550ML 50 mL
B99405500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B9940550ML Supplier Sigma Aldrich Supplier No. B9940550ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-tert-Butylphenol

Specifications

CAS 88-18-6
Melting Point -7°C (lit.)
Density 0.978 g/mL (at 25°C (literature))
Boiling Point 224°C (lit.)
Molecular Formula C10H14O
Refractive Index n20/D 1.523 (literature)
Linear Formula (CH3)3 CC6H4OH
MDL Number MFCD00002223
Quantity 50 mL
Molecular Weight (g/mol) 150.22
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.