accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88-53-9

CH3NH2ClSOHOO

CAS RN 88-53-9

IUPAC Name: 2-amino-5-chloro-4-methylbenzene-1-sulfonic acid
Synonyms: 2-amino-5-chloro-4-methylbenzenesulfonic acid
Molecular Weight (g/mol): 221.66
Molecular Formula: C7H8ClNO3S
InChi Key: VYZCFAPUHSSYCC-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(C=C1Cl)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results