Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-chlorotoluene-4-sulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. A0287500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0287500G 500 g
A028725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0287500G Supplier TCI America Supplier No. A0287500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88-53-9
Molecular Formula C7H8ClNO3S
Molecular Weight (g/mol) 221.655
MDL Number MFCD00043924
InChI Key VYZCFAPUHSSYCC-UHFFFAOYSA-N
Synonym 4-Chloro-5-methylaniline-2-sulfonic Acid, 4-Chloro-m-toluidine-6-sulfonic Acid, 2-Amino-5-chloro-4-methylbenzenesulfonic Acid
PubChem CID 6936
IUPAC Name 2-amino-5-chloro-4-methylbenzenesulfonic acid
SMILES CC1=CC(=C(C=C1Cl)S(=O)(=O)O)N

Specifications

Quantity 500 g
Formula Weight 221.66
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-chlorotoluene-4-sulfonic Acid
EINECSNumber (3)-2024
RTECSNumber DB4807000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.