accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNH2H2NSOOO

CAS RN 88-63-1

IUPAC Name: sodium 2,4-diaminobenzene-1-sulfonate
Synonyms: sodium 2,4-diaminobenzenesulfonate
Molecular Weight (g/mol): 210.18
Molecular Formula: C6H7N2NaO3S
InChi Key: WYSWTEPAYPNWDV-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC=C(C(N)=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results