accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88-68-6

IUPAC Name: 2-aminobenzamide
Synonyms: benzamide, 2-amino-
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: PXBFMLJZNCDSMP-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1N
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (4)
  • (5)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)