Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Anthranilamide

Catalog No. 501838918
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501838918 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501838918 Supplier Sigma Aldrich Supplier No. A898045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Anthranilamide
TRUSTED_SUSTAINABILITY

Specifications

CAS 88-68-6
Melting Point 111°C to 113°C (lit.)
Molecular Formula C7H8N2O
Linear Formula 2-(H2N)C6H4CONH2
MDL Number MFCD00007981
Quantity 1 g
Synonym 2-AB; 2-Aminobenzamide; Anthranilic acid amide
Molecular Weight (g/mol) 136.15
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.