accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(S)(R)O(R)NH

CAS RN 88056-92-2

IUPAC Name: (3R,5S,8aR)-5-cyano-3-phenyl-octahydro-[1,3]oxazolo[3,2-a]pyridin-4-ium
Synonyms: (3R,5S,8aR)-5-cyano-3-phenyl-octahydro-[1,3]oxazolo[3,2-a]pyridin-4-ium
Molecular Weight (g/mol): 229.3
Molecular Formula: C14H17N2O
InChi Key: GQHMNZGZXHZLEN-MELADBBJSA-O
SMILES: N#C[C@@H]1CCC[C@H]2OC[C@H]([NH+]12)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results