Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-2-Cyano-6-phenyloxazolopiperidine 98.0+%, TCI America™
SDP

Catalog No. C13631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13631G Supplier TCI America Supplier No. C13631G
 Restricted Item
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88056-92-2
Molecular Formula C14H17N2O
Molecular Weight (g/mol) 229.30
MDL Number MFCD00142338,MFCD08276761
InChI Key GQHMNZGZXHZLEN-MELADBBJSA-O
Synonym --2-cyano-6-phenyloxazolopiperidine, 2s,6r,9r-9-phenyl-7-oxa-1-azabicyclo 4.3.0 nonane-2-carbonitrile, 3r,5s,8ar-3-phenylhexahydro-2h-oxazolo 3,2-a pyridine-5-carbonitrile, 3r,5s,8ar---hexahydro-3-phenyl-5h-oxazolo 3,2-a pyridine-5-carbonitrile, 3r,5s,8ar-3-phenylhexahydro-5h-1,3 oxazolo 3,2-a pyridine-5-carbonitrile, 3r,5s,8ar-3-phenyl-hexahydro-2h-1,3 oxazolo 3,2-a pyridine-5-carbonitrile, --5-cyano-3-phenyl-2,3,5,6,7,8-hexahydro-5h-oxazolo 3,2-a pyridine, --2,3,5,6,7,8-hexahydro-3-phenyl-5h-oxazolo 3,2-a pyridine-5-carbonitrile, 8aalpha-3beta-phenylhexahydro-5h-oxazolo 3,2-a pyridine-5alpha-carbonitrile
PubChem CID 2724819
IUPAC Name (3R,5S,8aR)-5-cyano-3-phenyl-octahydro-[1,3]oxazolo[3,2-a]pyridin-4-ium
SMILES N#C[C@@H]1CCC[C@H]2OC[C@H]([NH+]12)C1=CC=CC=C1

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 1 g
Formula Weight 228.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-2-Cyano-6-phenyloxazolopiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.