CAS RN 881-86-7
IUPAC Name:
2,5-dimethyl pyridine-2,5-dicarboxylate
Synonyms:
2,5-dimethyl pyridine-2,5-dicarboxylate
Molecular Weight (g/mol):
195.17
Molecular Formula:
C9H9NO4
InChi Key:
TUGSJNQAIMFEDY-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(N=C1)C(=O)OC