Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylepyridine-2,5-dicarboxylate, 97%, Thermo Scientific™

Catalog No. RF02989EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RF02989EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF02989EA Supplier Thermo Scientific Chemicals Supplier No. RF02989EA
Add to Cart

Chemical Identifiers

CAS 881-86-7
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00034767
InChI Key TUGSJNQAIMFEDY-UHFFFAOYSA-N
Synonym dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate
PubChem CID 234911
IUPAC Name dimethyl pyridine-2,5-dicarboxylate
SMILES COC(=O)C1=CN=C(C=C1)C(=O)OC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 195.17
Percent Purity 97%
Chemical Name or Material Dimethylepyridine-2, 5-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.