accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 881005-91-0

IUPAC Name: 3,4,5-trihydroxy-6-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxane-2-carboxylic acid hydrate
Synonyms: 3,4,5-trihydroxy-6-[(4-methyl-2-oxochromen-7-yl)oxy]oxane-2-carboxylic acid hydrate
Molecular Weight (g/mol): 370.31
Molecular Formula: C16H18O10
InChi Key: URVSQZMOFUEQAW-UHFFFAOYNA-N
SMILES: O.CC1=CC(=O)OC2=CC(OC3OC(C(O)C(O)C3O)C(O)=O)=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results