accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88107-10-2

IUPAC Name: 1-{2-hydroxy-3-propyl-4-[4-(2H-1,2,3,4-tetrazol-5-yl)butoxy]phenyl}ethan-1-one
Synonyms: tomelukast tomelukastum
Molecular Weight (g/mol): 318.38
Molecular Formula: C16H22N4O3
InChi Key: MWYHLEQJTQJHSS-UHFFFAOYSA-N
SMILES: CCCC1=C(O)C(=CC=C1OCCCCC1=NNN=N1)C(C)=O
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results