accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOBrSF

CAS RN 88149-94-4

IUPAC Name: 5-bromo-2-(4-fluorophenyl)-3-(4-methanesulfonylphenyl)thiophene
Synonyms: C17H12BrFO2S2
Molecular Weight (g/mol): 411.3
Molecular Formula: C17H12BrFO2S2
InChi Key: AJFTZWGGHJXZOB-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)C1=C(SC(Br)=C1)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results