accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C(S)NHOOOHN(S)O

CAS RN 88191-84-8

IUPAC Name: benzyl N-[(1S)-2-methyl-1-{[(2S)-1-oxo-3-phenylpropan-2-yl]carbamoyl}propyl]carbamate
Synonyms: benzyl N-[(1S)-2-methyl-1-{[(2S)-1-oxo-3-phenylpropan-2-yl]carbamoyl}propyl]carbamate
Molecular Weight (g/mol): 382.46
Molecular Formula: C22H26N2O4
InChi Key: NGBKFLTYGSREKK-PMACEKPBSA-N
SMILES: CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)N[C@@H](CC1=CC=CC=C1)C=O
Product Type 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results