accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOCH3HOO

CAS RN 88210-96-2

IUPAC Name: 5,6-dimethoxy-1H-indole-2-carboxylic acid
Synonyms: 5,6-dimethoxy-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 221.21
Molecular Formula: C11H11NO4
InChi Key: UZHQHNDRVITJPL-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C=C(NC2=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)