accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3

CAS RN 88336-88-3

IUPAC Name: methyl 3-methyl-15-phenylpentadecanoate
Synonyms: methyl 3-methyl-15-phenylpentadecanoate
Molecular Weight (g/mol): 346.56
Molecular Formula: C23H38O2
InChi Key: JPSAJPNKLSJSSO-UHFFFAOYNA-N
SMILES: COC(=O)CC(C)CCCCCCCCCCCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results