Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Methyl-15-phenylpentadecanoate 95.0+%, TCI America™
SDP

Catalog No. M1407100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1407100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1407100MG Supplier TCI America Supplier No. M1407100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88336-88-3
Molecular Formula C23H38O2
Molecular Weight (g/mol) 346.56
MDL Number MFCD04038423
InChI Key JPSAJPNKLSJSSO-UHFFFAOYNA-N
Synonym beta-Methylbenzenepentadecanoic Acid Methyl Ester, Methyl beta-Methylbenzenepentadecanoate, 3-Methyl-15-phenylpentadecanoic Acid Methyl Ester
PubChem CID 13171669
IUPAC Name methyl 3-methyl-15-phenylpentadecanoate
SMILES COC(=O)CC(C)CCCCCCCCCCCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 205°C
Quantity 100 mg
Formula Weight 346.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Methyl-15-phenylpentadecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.