CAS RN 883731-58-6
IUPAC Name:
[4-(trifluoromethyl)cyclohexyl]methanol
Synonyms:
[4-(trifluoromethyl)cyclohexyl]methanol
Molecular Weight (g/mol):
182.19
Molecular Formula:
C8H13F3O
InChi Key:
DYUAPKHFRXRMTL-UHFFFAOYSA-N
SMILES:
OCC1CCC(CC1)C(F)(F)F
1
–
1
of
1
results
1
4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture) 98.0+%, TCI America™
CAS: 883731-58-6 Molecular Formula: C8H13F3O Molecular Weight (g/mol): 182.19 MDL Number: MFCD21324328,MFCD29054589 InChI Key: DYUAPKHFRXRMTL-UHFFFAOYSA-N PubChem CID: 44228198 IUPAC Name: [4-(trifluoromethyl)cyclohexyl]methanol SMILES: OCC1CCC(CC1)C(F)(F)F