Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. T28851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28851G Supplier TCI America Supplier No. T28851G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 883731-58-6
Molecular Formula C8H13F3O
Molecular Weight (g/mol) 182.19
MDL Number MFCD21324328,MFCD29054589
InChI Key DYUAPKHFRXRMTL-UHFFFAOYSA-N
PubChem CID 44228198
IUPAC Name [4-(trifluoromethyl)cyclohexyl]methanol
SMILES OCC1CCC(CC1)C(F)(F)F

Specifications

Color Colorless
Quantity 1 g
Formula Weight 182.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.