accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNNCH3

CAS RN 883738-27-0

IUPAC Name: 1-methyl-4-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperazine
Synonyms: 1-methyl-4-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperazine
Molecular Weight (g/mol): 316.25
Molecular Formula: C18H29BN2O2
InChi Key: PESZAXRJFGLGTR-UHFFFAOYSA-N
SMILES: CN1CCN(CC2=CC=CC(=C2)B2OC(C)(C)C(C)(C)O2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results