accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNOOHO

CAS RN 88374-37-2

IUPAC Name: [1-(5-nitropyridin-2-yl)pyrrolidin-2-yl]methanol
Synonyms: [1-(5-nitropyridin-2-yl)pyrrolidin-2-yl]methanol
Molecular Weight (g/mol): 223.23
Molecular Formula: C10H13N3O3
InChi Key: YBARIBJRNJYVTK-UHFFFAOYNA-N
SMILES: OCC1CCCN1C1=NC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results