Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-N-(5-Nitro-2-pyridyl)prolinol 99.0+%, TCI America™
SDP

Catalog No. N05401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05401G Supplier TCI America Supplier No. N05401G

Specifications

CAS 88374-37-2
Melting Point 83°C
Color Yellow
Molecular Formula C10H13N3O3
Quantity 1 g
MDL Number MFCD00145250
Synonym (S)-(-)-1-(5-Nitro-2-pyridyl)-2-pyrrolidinemethanol, 2-(N-Prolinol)-5-nitropyridine
InChI Key YBARIBJRNJYVTK-UHFFFAOYNA-N
SMILES OCC1CCCN1C1=NC=C(C=C1)[N+]([O-])=O
IUPAC Name [1-(5-nitropyridin-2-yl)pyrrolidin-2-yl]methanol
Molecular Weight (g/mol) 223.23
PubChem CID 44630404
Formula Weight 223.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-N-(5-Nitro-2-pyridyl)prolinol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.