accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClO

CAS RN 88419-56-1

IUPAC Name: 2,4,5-trifluorobenzoyl chloride
Synonyms: 2,4,5-trifluorobenzoyl chloride
Molecular Weight (g/mol): 194.54
Molecular Formula: C7H2ClF3O
InChi Key: STBGCAUUOPNJBH-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C=C1F)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results