Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluorobenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. T16535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16535G Supplier TCI America Supplier No. T16535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88419-56-1
Molecular Formula C7H2ClF3O
Molecular Weight (g/mol) 194.54
MDL Number MFCD00061204
InChI Key STBGCAUUOPNJBH-UHFFFAOYSA-N
Synonym 2,4,5-trifluorobenzoylchloride, benzoyl chloride, 2,4,5-trifluoro, 2,4,5-tris fluoranyl benzoyl chloride, pubchem9716, acmc-209qt0, 2,4,5-trifluorobenzoyl-chloride, stbgcauuopnjbh-uhfffaoysa, timtec-bb sbb006657, 2,4,5-trifluoro-benzoyl chloride, 2,4,5-trifluorobenzoyl chloride
PubChem CID 145164
IUPAC Name 2,4,5-trifluorobenzoyl chloride
SMILES FC1=CC(F)=C(C=C1F)C(Cl)=O

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 3265
Formula Weight 194.54
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trifluorobenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.