accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONFBr

CAS RN 884494-81-9

IUPAC Name: 3-bromo-5-fluoro-2-methoxypyridine
Synonyms: 3-bromo-5-fluoro-2-methoxypyridine
Molecular Weight (g/mol): 206.01
Molecular Formula: C6H5BrFNO
InChi Key: HSWCOAGQYSBFAK-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(F)C=N1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)