Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-fluoro-2-methoxypyridine 97.0+%, TCI America™
SDP

Catalog No. B51341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B51341G Supplier TCI America Supplier No. B51341G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorinated pyridine building block with ≥97.0% purity.

Chemical Identifiers

CAS 884494-81-9
Molecular Formula C6H5BrFNO
Molecular Weight (g/mol) 206.014
MDL Number MFCD04972397
InChI Key HSWCOAGQYSBFAK-UHFFFAOYSA-N
Synonym 3-bromo-5-fluoro-2-methoxypyrdine, 2-methoxy-3-bromo-5-fluoropyridine, synquest 4h07-b-01, 3-bromo-5-fluoro-2-methoxy-pyridine, pyridine,3-bromo-5-fluoro-2-methoxy, pyridine, 3-bromo-5-fluoro-2-methoxy, pubchem1175, acmc-209qt8, 3-bromo-5-fluoropyridin-2-yl methyl ether, 3-bromo-5-fluoro-2-methoxy pyridine
PubChem CID 40427006
IUPAC Name 3-bromo-5-fluoro-2-methoxypyridine
SMILES COC1=C(C=C(C=N1)F)Br

Specifications

Quantity 1 g
Formula Weight 206.01
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-fluoro-2-methoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.