accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CN

CAS RN 884507-39-5

IUPAC Name: 1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}pyrrolidine
Synonyms: 1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}pyrrolidine
Molecular Weight (g/mol): 287.21
Molecular Formula: C17H26BNO2
InChi Key: AGSIDMRVRGPBIE-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results