accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 884530-69-2

IUPAC Name: N,N-bis(4-bromophenyl)-[1,1'-biphenyl]-4-amine
Synonyms: N,N-bis(4-bromophenyl)-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 479.22
Molecular Formula: C24H17Br2N
InChi Key: BKJULDLPGWGCHY-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N(C1=CC=C(Br)C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results