Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibromo-4″-phenyltriphenylamine 97.0+%, TCI America™
SDP

Catalog No. D33031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33031G Supplier TCI America Supplier No. D33031G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 884530-69-2
Molecular Formula C24H17Br2N
Molecular Weight (g/mol) 479.215
InChI Key BKJULDLPGWGCHY-UHFFFAOYSA-N
Synonym N-(4-Biphenylyl)-N,N-bis(4-bromophenyl)amine
PubChem CID 22709142
IUPAC Name N,N-bis(4-bromophenyl)-4-phenylaniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br

Specifications

Melting Point 138°C
Color White
Quantity 1 g
Formula Weight 479.22
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromo-4′′-phenyltriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.