CAS RN 88496-70-2
IUPAC Name:
methyl 4-chloro-3-hydroxybutanoate
Synonyms:
methyl 4-chloro-3-hydroxybutanoate
Molecular Weight (g/mol):
152.57
Molecular Formula:
C5H9ClO3
InChi Key:
WMRINGSAVOPXTE-UHFFFAOYNA-N
SMILES:
COC(=O)CC(O)CCl
1
–
1
of
1
results
1
Methyl (R)-4-Chloro-3-hydroxybutyrate, TCI America™
CAS: 88496-70-2 Molecular Formula: C5H9ClO3 Molecular Weight (g/mol): 152.574 MDL Number: MFCD00273367 InChI Key: WMRINGSAVOPXTE-SCSAIBSYSA-N Synonym: (R)-4-Chloro-3-hydroxybutyric Acid Methyl Ester PubChem CID: 10920707 IUPAC Name: methyl (3R)-4-chloro-3-hydroxybutanoate SMILES: COC(=O)CC(CCl)O