Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (R)-4-Chloro-3-hydroxybutyrate, TCI America™
SDP

Catalog No. M14835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14835G Supplier TCI America Supplier No. M14835G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88496-70-2
Molecular Formula C5H9ClO3
Molecular Weight (g/mol) 152.574
MDL Number MFCD00273367
InChI Key WMRINGSAVOPXTE-SCSAIBSYSA-N
Synonym (R)-4-Chloro-3-hydroxybutyric Acid Methyl Ester
PubChem CID 10920707
IUPAC Name methyl (3R)-4-chloro-3-hydroxybutanoate
SMILES COC(=O)CC(CCl)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 152.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (R)-4-Chloro-3-hydroxybutyrate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.