accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFNHN

CAS RN 885271-64-7

IUPAC Name: 5-(trifluoromethyl)-1H-indazole
Synonyms: 5-(trifluoromethyl)-1H-indazole
Molecular Weight (g/mol): 186.14
Molecular Formula: C8H5F3N2
InChi Key: WBHMPANPIGWXQV-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C2NN=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results