Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Trifluoromethyl)-1H-indazole, 97%, Thermo Scientific™

Catalog No. MAY00296DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAY00296DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00296DA Supplier Thermo Scientific Chemicals Supplier No. MAY00296DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 885271-64-7
Molecular Formula C8H5F3N2
Molecular Weight (g/mol) 186.137
InChI Key WBHMPANPIGWXQV-UHFFFAOYSA-N
Synonym 5-trifluoromethyl-1h-indazole, 1h-indazole,5-trifluoromethyl
PubChem CID 21409222
IUPAC Name 5-(trifluoromethyl)-1H-indazole
SMILES C1=CC2=C(C=C1C(F)(F)F)C=NN2

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 186.14
Chemical Name or Material 5-(Trifluoromethyl)-1H-indazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.