accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88535-47-1

IUPAC Name: 4-nitro-1,2,3,6,7,8-hexahydropyrene
Synonyms: 4-nitro-1,2,3,6,7,8-hexahydropyrene
Molecular Weight (g/mol): 253.3
Molecular Formula: C16H15NO2
InChi Key: KVZSMBRWPWEQHJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C3C(CCC2)=CC=C2CCCC1=C32
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results