Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-1,2,3,6,7,8-hexahydropyrene 98.0+%, TCI America™
SDP

Catalog No. N10611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10611G Supplier TCI America Supplier No. N10611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88535-47-1
Molecular Formula C16H15NO2
Molecular Weight (g/mol) 253.301
MDL Number MFCD27964325
InChI Key KVZSMBRWPWEQHJ-UHFFFAOYSA-N
PubChem CID 184879
IUPAC Name 4-nitro-1,2,3,6,7,8-hexahydropyrene
SMILES C1CC2=C3C(=CC(=C4C3=C(CCC4)C=C2)[N+](=O)[O-])C1

Specifications

Melting Point 92°C
Color Yellow
Quantity 1 g
Formula Weight 253.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-1,2,3,6,7,8-hexahydropyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.