accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3H3CCH3NNH3C

CAS RN 885698-95-3

IUPAC Name: 2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole
Synonyms: 2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole
Molecular Weight (g/mol): 258.13
Molecular Formula: C14H19BN2O2
InChi Key: YQMBOMPXHDRRIM-UHFFFAOYSA-N
SMILES: CN1C=C2C(C=CC=C2B2OC(C)(C)C(C)(C)O2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results