accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88574-07-6

OOOHONH

CAS RN 88574-07-6

IUPAC Name: 11-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)undecanoic acid
Synonyms: 11-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}undecanoic acid
Molecular Weight (g/mol): 423.55
Molecular Formula: C26H33NO4
InChi Key: IMEVDILDSCXKJW-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCNC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)