accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNOBr

CAS RN 885952-25-0

IUPAC Name: N-benzyl-4-(benzyloxy)-5-bromopyrimidin-2-amine
Synonyms: N-benzyl-4-(benzyloxy)-5-bromopyrimidin-2-amine
Molecular Weight (g/mol): 370.25
Molecular Formula: C18H16BrN3O
InChi Key: OWAQGZVVTHQXOM-UHFFFAOYSA-N
SMILES: BrC1=CN=C(NCC2=CC=CC=C2)N=C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results