accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNNHCH3CH3H3CNSH3CN

CAS RN 886-50-0

IUPAC Name: N2-tert-butyl-N4-ethyl-6-(methylsulfanyl)-1,3,5-triazine-2,4-diamine
Synonyms: shortstop igran epa pesticide chemical code 080813 short-stop E igran 500 terbutryn terbutrin terbutrex caswell no. 125D clarosan
Molecular Weight (g/mol): 241.36
Molecular Formula: C10H19N5S
InChi Key: IROINLKCQGIITA-UHFFFAOYSA-N
SMILES: CCNC1=NC(NC(C)(C)C)=NC(SC)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results