accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886364-22-3

IUPAC Name: 3-methyl-4-nitrobenzene-1-carbothioamide
Synonyms: 3-methyl-4-nitrobenzenecarbothioamide
Molecular Weight (g/mol): 196.22
Molecular Formula: C8H8N2O2S
InChi Key: QNPFIXDGSZDAGC-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(=C1)C(N)=S)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results