accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHOBr

CAS RN 886365-43-1

IUPAC Name: 5-bromo-3-methylpyridine-2-carboxylic acid
Synonyms: 5-bromo-3-methylpyridine-2-carboxylic acid
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: GSOCTKHBFREQCV-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CN=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results