accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONH2

CAS RN 88675-24-5

IUPAC Name: oxolan-3-amine
Synonyms: oxolan-3-amine
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: MIPHRQMEIYLZFZ-UHFFFAOYNA-N
SMILES: NC1CCOC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results