accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFIF

CAS RN 886762-68-1

IUPAC Name: 1,3-difluoro-5-iodo-2-methoxybenzene
Synonyms: 1,3-difluoro-5-iodo-2-methoxybenzene
Molecular Weight (g/mol): 270.02
Molecular Formula: C7H5F2IO
InChi Key: VPBVKAYYQNPEGB-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(I)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results