Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoro-4-iodoanisole, 99%, Thermo Scientific™

Catalog No. AAH3160706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3160706 5 g
AAH3160703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3160706 Supplier Thermo Scientific Chemicals Supplier No. H3160706

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886762-68-1
Molecular Formula C7H5F2IO
Molecular Weight (g/mol) 270.02
MDL Number MFCD07368787
InChI Key VPBVKAYYQNPEGB-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-iodoanisole, benzene,1,3-difluoro-5-iodo-2-methoxy, 1,3-difluoro-2-methoxy-5-iodobenzene
PubChem CID 2783151
IUPAC Name 1,3-difluoro-5-iodo-2-methoxybenzene
SMILES COC1=C(F)C=C(I)C=C1F

Specifications

Refractive Index 1.556
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 270.02
Percent Purity 99%
Chemical Name or Material 2,6-Difluoro-4-iodoanisole
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.